The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152450.2 | NP_689663.2 | 5 - 6 | 728 | 83 |
XM_005254166.2 | XP_005254223.1 | 5 - 6 | 641 | 83 | |
XM_006720398.3 | XP_006720461.1 | 7 - 8 | 910 | 83 | |
XM_006720399.2 | XP_006720462.1 | 5 - 6 | 701 | 83 | |
XM_011521247.2 | XP_011519549.1 | 7 - 8 | 874 | 83 | |
XM_011521248.2 | XP_011519550.1 | 6 - 7 | 755 | 83 | |
XM_011521249.2 | XP_011519551.1 | 6 - 7 | 865 | 83 | |
XM_011521250.2 | XP_011519552.1 | 6 - 7 | 718 | 83 | |
XM_017021931.1 | XP_016877420.1 | 6 - 7 | 1166 | 83 | |
XM_017021932.1 | XP_016877421.1 | 5 - 6 | 756 | 83 | |
GenBank mRNA | AK307975.1 | - | 5 - 6 | 671 | 83 |
BC022537.1 | - | 5 - 6 | 728 | 83 |