The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002432.1 | NP_002423.1 | 3 - 4 | 597 | 68 |
GenBank mRNA | AK290392.1 | - | 3 - 4 | 604 | 68 |
AK313210.1 | - | 3 - 4 | 608 | 68 | |
BC020715.1 | - | 3 - 4 | 635 | 68 | |
BC032319.1 | - | 3 - 4 | 608 | 68 | |
M81750.1 | - | 3 - 4 | 597 | 68 |