The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005038.2 | NP_005029.1 | 4 - 5 | 638 | 101 |
GenBank mRNA | AK313929.1 | - | 4 - 5 | 639 | 101 |
BC030707.2 | - | 4 - 5 | 634 | 101 | |
CR542081.1 | - | 4 - 5 | 526 | 101 | |
L11667.1 | - | 4 - 5 | 625 | 101 |