Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAPRE2
Entrez Gene ID
10982
Gene Name
microtubule associated protein RP/EB family member 2
Gene Aliases
CSCSC2, EB1, EB2, RP1
Chromosome Location
Chr.18: 34976928 - 35143470 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143826.2 NP_001137298.1 3 - 4 341 108
NM_001143827.2 NP_001137299.1 3 - 4 487 108
NM_014268.3 NP_055083.1 2 - 3 429 108
NR_046177.1 2 - 3 244 108
GenBank mRNA AK294833.1 - 3 - 4 431 108
AK296251.1 - 2 - 3 244 108
AK315766.1 - 2 - 3 382 108
BC007318.2 - 2 - 3 372 108
BT020086.1 - 2 - 3 249 108
CR536545.1 - 2 - 3 249 108
DC335558.1 - 3 - 4 489 108
DC341673.1 - 3 - 4 341 108
DC354326.1 - 2 - 3 429 108
X94232.1 - 2 - 3 389 108

View Full Product Details