The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143826.2 | NP_001137298.1 | 7 - 8 | 1004 | 72 |
NM_001143827.2 | NP_001137299.1 | 7 - 8 | 1150 | 72 | |
NM_001256420.1 | NP_001243349.1 | 5 - 6 | 964 | 72 | |
NM_014268.3 | NP_055083.1 | 6 - 7 | 1092 | 72 | |
NR_046177.1 | 6 - 7 | 907 | 72 | ||
GenBank mRNA | AK090945.1 | - | 5 - 6 | 920 | 72 |
AK294833.1 | - | 7 - 8 | 1094 | 72 | |
AK296251.1 | - | 6 - 7 | 907 | 72 | |
AK315766.1 | - | 6 - 7 | 1045 | 72 | |
BC007318.2 | - | 6 - 7 | 1035 | 72 | |
BT020086.1 | - | 6 - 7 | 912 | 72 | |
CR536545.1 | - | 6 - 7 | 912 | 72 | |
X94232.1 | - | 6 - 7 | 1052 | 72 |