The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172643.1 | NP_001166114.1 | 4 - 5 | 202 | 69 |
NM_001172644.1 | NP_001166115.1 | 3 - 4 | 563 | 69 | |
NM_001271934.1 | NP_001258863.1 | 5 - 6 | 733 | 69 | |
NM_001271935.1 | NP_001258864.1 | 4 - 5 | 202 | 69 | |
NM_001272050.1 | NP_001258979.1 | 4 - 5 | 412 | 69 | |
NM_004614.4 | NP_004605.4 | 4 - 5 | 638 | 69 | |
NR_073520.1 | 3 - 4 | 1566 | 69 | ||
GenBank mRNA | AF521891.1 | - | 4 - 5 | 638 | 69 |
AK302976.1 | - | 4 - 5 | 416 | 69 | |
AK316226.1 | - | 3 - 4 | 273 | 69 | |
BC113928.1 | - | 4 - 5 | 181 | 69 | |
BC134344.1 | - | 4 - 5 | 181 | 69 | |
BC142970.1 | - | 5 - 6 | 534 | 69 | |
DA500158.1 | - | 5 - 6 | 328 | 69 | |
U77088.1 | - | 4 - 5 | 202 | 69 | |
U80628.1 | - | 3 - 4 | 953 | 69 | |
Y10498.3 | - | 4 - 5 | 403 | 69 |