Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC20B
Entrez Gene ID
166979
Gene Name
cell division cycle 20B
Gene Aliases
G6VTS76519
Chromosome Location
Chr.5: 55112971 - 55173177 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170402.1 NP_001163873.1 9 - 10 1390 58
NM_152623.2 NP_689836.2 9 - 10 1390 58
GenBank mRNA AB077043.1 - 9 - 10 1359 58
AB081750.1 - 9 - 10 1359 58
AK095246.1 - 9 - 10 1388 58
BC136572.1 - 9 - 10 1260 58

View Full Product Details