The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176813.4 | NP_789783.1 | 4 - 5 | 302 | 154 |
XM_005249627.2 | XP_005249684.1 | 4 - 5 | 281 | 154 | |
XM_011515152.2 | XP_011513454.1 | 4 - 5 | 294 | 154 | |
GenBank mRNA | AY069977.1 | - | 3 - 4 | 267 | 154 |
AY359065.1 | - | 4 - 5 | 286 | 154 | |
BC058284.1 | - | 4 - 5 | 307 | 154 | |
BM984550.1 | - | 4 - 5 | 154 | ||
BU682778.1 | - | 4 - 5 | 154 | ||
CB242846.1 | - | 4 - 5 | 154 |