The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012347.4 | NP_036479.1 | 10 - 11 | 1131 | 99 |
NM_033480.2 | NP_258441.1 | 11 - 12 | 1381 | 99 | |
NM_033481.3 | NP_258442.2 | 11 - 12 | 1258 | 99 | |
XM_005248995.4 | XP_005249052.1 | 11 - 12 | 1065 | 99 | |
XM_005248996.4 | XP_005249053.1 | 11 - 12 | 1216 | 99 | |
GenBank mRNA | AB209833.1 | - | 11 - 12 | 1420 | 99 |
AF174597.1 | - | 7 - 8 | 720 | 99 | |
AF176704.1 | - | 10 - 11 | 1131 | 99 | |
AK021559.1 | - | 11 - 12 | 1417 | 99 | |
AL137520.1 | - | 11 - 12 | 1381 | 99 | |
AY858798.1 | - | 11 - 12 | 1423 | 99 | |
BC000650.2 | - | 11 - 12 | 1258 | 99 | |
BC024973.2 | - | 8 - 9 | 802 | 99 | |
BX641082.1 | - | 3 - 4 | 1529 | 99 |