Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKRF
Entrez Gene ID
55922
Gene Name
NFKB repressing factor
Gene Aliases
ITBA4, NRF
Chromosome Location
Chr.X: 119588337 - 119606464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173487.1 NP_001166958.1 1 - 2 149 62
GenBank mRNA AL539002.3 - 1 - 2 78 62

View Full Product Details