The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173487.1 | NP_001166958.1 | 2 - 3 | 254 | 107 |
GenBank mRNA | AL539002.3 | - | 2 - 3 | 183 | 107 |