Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STMN4
Entrez Gene ID
81551
Gene Name
stathmin 4
Gene Aliases
RB3
Chromosome Location
Chr.8: 27235323 - 27258865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283053.1 NP_001269982.1 1 - 2 87 87
NM_001283054.1 NP_001269983.1 1 - 2 87 87
NM_030795.3 NP_110422.2 1 - 2 87 87
XM_005273652.4 XP_005273709.1 1 - 2 513 87
XM_005273655.4 XP_005273712.1 1 - 2 513 87
GenBank mRNA AK225142.1 - 1 - 2 53 87
AK294023.1 - 1 - 2 52 87
AK297415.1 - 1 - 2 68 87
AK309207.1 - 1 - 2 52 87
AL136568.1 - 1 - 2 57 87

View Full Product Details