The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001283053.1 | NP_001269982.1 | 6 - 7 | 759 | 61 |
NM_001283055.1 | NP_001269984.1 | 4 - 5 | 587 | 61 | |
XM_005273655.4 | XP_005273712.1 | 5 - 6 | 1104 | 61 | |
GenBank mRNA | AK295329.1 | - | 4 - 5 | 549 | 61 |
AK297415.1 | - | 6 - 7 | 740 | 61 |