Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STMN4
Entrez Gene ID
81551
Gene Name
stathmin 4
Gene Aliases
RB3
Chromosome Location
Chr.8: 27235323 - 27258865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283053.1 NP_001269982.1 7 - 8 817 85
NM_001283055.1 NP_001269984.1 5 - 6 645 85
XM_005273655.4 XP_005273712.1 6 - 7 1162 85
GenBank mRNA AK295329.1 - 5 - 6 607 85
AK297415.1 - 7 - 8 798 85

View Full Product Details