The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290104.1 | NP_001277033.1 | 15 - 16 | 2149 | 86 |
NM_003460.2 | NP_003451.1 | 15 - 16 | 2176 | 86 | |
XM_011545942.2 | XP_011544244.1 | 14 - 15 | 1705 | 86 | |
GenBank mRNA | AK310454.1 | - | 15 - 16 | 2176 | 86 |
AK313003.1 | - | 14 - 15 | 1718 | 86 | |
BC096304.3 | - | 14 - 15 | 1693 | 86 | |
BC096305.3 | - | 14 - 15 | 1666 | 86 | |
BC096306.1 | - | 14 - 15 | 1693 | 86 | |
BC096307.3 | - | 14 - 15 | 1693 | 86 | |
M90366.1 | - | 14 - 15 | 1693 | 86 |