The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033481.3 | NP_258442.2 | 1 - 2 | 216 | 109 |
GenBank mRNA | BC000650.2 | - | 1 - 2 | 216 | 109 |