The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005921.1 | NP_005912.1 | 18 - 19 | 4755 | 94 |
XM_017009484.1 | XP_016864973.1 | 18 - 19 | 7466 | 94 | |
XM_017009485.1 | XP_016864974.1 | 18 - 19 | 4194 | 94 | |
GenBank mRNA | AF042838.1 | - | 18 - 19 | 4203 | 94 |
BU194120.1 | - | 5 - 6 | 714 | 94 | |
DA889202.1 | - | 2 - 3 | 160 | 94 |