The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005921.1 | NP_005912.1 | 3 - 4 | 1334 | 118 |
XM_017009484.1 | XP_016864973.1 | 3 - 4 | 4045 | 118 | |
XM_017009485.1 | XP_016864974.1 | 3 - 4 | 773 | 118 | |
GenBank mRNA | AF042838.1 | - | 3 - 4 | 776 | 118 |
AK307828.1 | - | 4 - 5 | 554 | 118 |