The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004776.3 | NP_004767.1 | 8 - 9 | 1214 | 66 |
GenBank mRNA | AB004550.1 | - | 8 - 9 | 1132 | 66 |
AB871482.2 | - | 8 - 9 | 1013 | 66 | |
AF038663.1 | - | 8 - 9 | 1030 | 66 | |
BC074821.2 | - | 8 - 9 | 1053 | 66 | |
BC074873.2 | - | 8 - 9 | 1053 | 66 | |
BC104987.1 | - | 8 - 9 | 1053 | 66 | |
BC112265.1 | - | 8 - 9 | 1053 | 66 |