The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000615.6 | NP_000606.3 | 17 - 18 | 2674 | 68 |
NM_001242607.1 | NP_001229536.1 | 19 - 20 | 2782 | 68 | |
NM_181351.4 | NP_851996.2 | 18 - 19 | 2704 | 68 | |
GenBank mRNA | AK057509.1 | - | 17 - 18 | 2777 | 68 |
AK292453.1 | - | 19 - 20 | 2339 | 68 | |
AK299865.1 | - | 8 - 9 | 885 | 68 | |
AK314589.1 | - | 19 - 20 | 2754 | 68 | |
BC014205.2 | - | 12 - 13 | 1568 | 68 | |
BC047244.1 | - | 18 - 19 | 2516 | 68 | |
M17410.1 | - | 6 - 7 | 829 | 68 | |
S71824.1 | - | 17 - 18 | 2509 | 68 | |
U63041.1 | - | 17 - 18 | 2345 | 68 |