Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
Chromosome Location
Chr.11: 112961247 - 113278436 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076682.3 NP_001070150.1 15 - 16 2218 62
GenBank mRNA AK303232.1 - 7 - 8 487 62
M17409.1 - 5 - 6 382 62
M22094.1 - 5 - 6 382 62
X16841.1 - 15 - 16 1997 62

View Full Product Details