Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NADK
Entrez Gene ID
65220
Gene Name
NAD kinase
Gene Aliases
dJ283E3.1
Chromosome Location
Chr.1: 1751232 - 1780514 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198993.1 NP_001185922.1 4 - 5 545 86
NM_001198995.1 NP_001185924.1 2 - 3 407 86
NM_023018.4 NP_075394.3 4 - 5 614 86
XM_005244778.2 XP_005244835.1 4 - 5 675 86
XM_006710837.2 XP_006710900.1 5 - 6 790 86
XM_006710838.2 XP_006710901.1 4 - 5 523 86
XM_006710839.1 XP_006710902.1 7 - 8 1102 86
XM_017002143.1 XP_016857632.1 4 - 5 675 86
XM_017002144.1 XP_016857633.1 2 - 3 419 86
GenBank mRNA AK023114.1 - 4 - 5 614 86
AK290161.1 - 4 - 5 596 86
AK291599.1 - 4 - 5 538 86
AK296722.1 - 2 - 3 407 86
AY090771.1 - 3 - 4 392 86
BC001709.1 - 4 - 5 477 86

View Full Product Details