Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NADK
Entrez Gene ID
65220
Gene Name
NAD kinase
Gene Aliases
dJ283E3.1
Chromosome Location
Chr.1: 1751232 - 1780514 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198993.1 NP_001185922.1 9 - 10 1094 84
NM_001198994.1 NP_001185923.1 11 - 12 1581 84
NM_001198995.1 NP_001185924.1 7 - 8 956 84
NM_023018.4 NP_075394.3 9 - 10 1163 84
XM_005244778.2 XP_005244835.1 9 - 10 1224 84
XM_006710837.2 XP_006710900.1 10 - 11 1339 84
XM_006710838.2 XP_006710901.1 9 - 10 1072 84
XM_006710839.1 XP_006710902.1 12 - 13 1651 84
XM_011542006.1 XP_011540308.1 8 - 9 1039 84
XM_011542007.1 XP_011540309.1 8 - 9 1118 84
XM_011542008.1 XP_011540310.1 8 - 9 976 84
XM_011542009.2 XP_011540311.1 7 - 8 867 84
XM_017002143.1 XP_016857632.1 9 - 10 1224 84
XM_017002144.1 XP_016857633.1 7 - 8 968 84
XM_017002145.1 XP_016857634.1 9 - 10 1407 84
XM_017002146.1 XP_016857635.1 9 - 10 1277 84
GenBank mRNA AF250320.1 - 11 - 12 1581 84
AK023114.1 - 9 - 10 1163 84
AK290161.1 - 9 - 10 1145 84
AK291599.1 - 9 - 10 1087 84
AK291728.1 - 9 - 10 1063 84
AK296722.1 - 7 - 8 956 84
AY090771.1 - 8 - 9 941 84
BC001709.1 - 9 - 10 1026 84
BC024219.1 - 2 - 3 251 84

View Full Product Details