Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFX6
Entrez Gene ID
222546
Gene Name
regulatory factor X6
Gene Aliases
MTCHRS, MTFS, RFXDC1, dJ955L16.1
Chromosome Location
Chr.6: 116877213 - 116932163 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173560.3 NP_775831.2 9 - 10 1027 112
XM_011535589.1 XP_011533891.1 8 - 9 914 112
XM_017010477.1 XP_016865966.1 8 - 9 834 112
GenBank mRNA BC039248.1 - 9 - 10 1027 112

View Full Product Details