The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006734.3 | NP_006725.3 | 6 - 7 | 6077 | 85 |
XM_017010805.1 | XP_016866294.1 | 6 - 7 | 6042 | 85 | |
GenBank mRNA | AK226163.1 | - | 2 - 3 | 3555 | 85 |
AK296531.1 | - | 2 - 3 | 325 | 85 | |
CR936782.1 | - | 6 - 7 | 6077 | 85 | |
DQ231041.1 | - | 2 - 3 | 5334 | 85 | |
X65644.1 | - | 2 - 3 | 5511 | 85 |