The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320336.1 | NP_001307265.1 | 5 - 6 | 1970 | 86 |
NM_005099.5 | NP_005090.3 | 5 - 6 | 1970 | 86 | |
GenBank mRNA | AB014588.1 | - | 5 - 6 | 1944 | 86 |
AF148213.1 | - | 5 - 6 | 1950 | 86 | |
AK291573.1 | - | 5 - 6 | 1969 | 86 | |
AY358886.1 | - | 5 - 6 | 1962 | 86 | |
BC063293.1 | - | 5 - 6 | 1969 | 86 |