Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMK2D
Entrez Gene ID
817
Gene Name
calcium/calmodulin dependent protein kinase II delta
Gene Aliases
CAMKD
Chromosome Location
Chr.4: 113451032 - 113761927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001221.3 NP_001212.2 14 - 15 1894 105
NM_001321574.1 NP_001308503.1 14 - 15 1891 105
NM_001321576.1 NP_001308505.1 14 - 15 1891 105
NM_172115.2 NP_742113.1 14 - 15 1894 105
NM_172127.2 NP_742125.1 14 - 15 1894 105
NM_172128.2 NP_742126.1 14 - 15 1894 105
XM_006714330.2 XP_006714393.1 14 - 15 1733 105
XM_017008678.1 XP_016864167.1 14 - 15 1709 105
GenBank mRNA AF071569.1 - 14 - 15 1035 105
AK025368.1 - 3 - 4 145 105
AK055642.1 - 14 - 15 1412 105
BC032784.1 - 14 - 15 1097 105
U50361.1 - 3 - 4 132 105

View Full Product Details