Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC61A2
Entrez Gene ID
55176
Gene Name
Sec61 translocon alpha 2 subunit
Gene Aliases
-
Chromosome Location
Chr.10: 12129641 - 12169958 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142627.2 NP_001136099.1 7 - 8 762 76
NM_001142628.1 NP_001136100.1 6 - 7 696 76
NM_018144.3 NP_060614.2 7 - 8 762 76
NR_024576.2 7 - 8 762 76
NR_024577.2 8 - 9 895 76
GenBank mRNA AF346603.1 - 7 - 8 703 76
AK022027.1 - 1 - 2 145 76
AK022640.1 - 7 - 8 697 76
AK057532.1 - 8 - 9 836 76
AK292295.1 - 7 - 8 712 76
AK301841.1 - 6 - 7 649 76
BC026179.1 - 7 - 8 712 76

View Full Product Details