The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020678.3 | NP_065729.1 | 1 - 2 | 174 | 85 |
GenBank mRNA | AF225421.1 | - | 1 - 2 | 145 | 85 |