The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322464.1 | NP_001309393.1 | 1 - 2 | 456 | 57 |
NM_030645.1 | NP_085148.1 | 1 - 2 | 126 | 57 | |
GenBank mRNA | AK056382.1 | - | 1 - 2 | 126 | 57 |
AK125138.1 | - | 1 - 2 | 803 | 57 | |
AY358453.1 | - | 1 - 2 | 48 | 57 |