Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMK2D
Entrez Gene ID
817
Gene Name
calcium/calmodulin dependent protein kinase II delta
Gene Aliases
CAMKD
Chromosome Location
Chr.4: 113451032 - 113761927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321567.1 NP_001308496.1 14 - 15 1906 71
NM_001321569.1 NP_001308498.1 14 - 15 1906 71
NM_001321570.1 NP_001308499.1 14 - 15 1906 71
NM_001321571.1 NP_001308500.1 14 - 15 1906 71
NM_001321572.1 NP_001308501.1 14 - 15 1906 71
NM_001321579.1 NP_001308508.1 14 - 15 1906 71
NM_001321586.1 NP_001308515.1 14 - 15 1903 71
XM_005263253.3 XP_005263310.1 14 - 15 1721 71
XM_005263254.3 XP_005263311.1 14 - 15 1742 71
XM_011532292.2 XP_011530594.1 10 - 11 756 71
XM_017008674.1 XP_016864163.1 14 - 15 1742 71
XM_017008675.1 XP_016864164.1 15 - 16 1846 71
XM_017008676.1 XP_016864165.1 14 - 15 1082 71
GenBank mRNA AK294517.1 - 14 - 15 1197 71
EF139846.1 - 3 - 4 132 71

View Full Product Details