The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014146.3 | NP_054865.2 | 6 - 7 | 631 | 76 |
NM_032463.2 | NP_115852.1 | 7 - 8 | 750 | 76 | |
NM_032464.2 | NP_115853.2 | 7 - 8 | 820 | 76 | |
XM_011516558.2 | XP_011514860.1 | 7 - 8 | 660 | 76 | |
GenBank mRNA | AF252611.1 | - | 7 - 8 | 571 | 76 |
AF252612.1 | - | 6 - 7 | 382 | 76 | |
AF252613.1 | - | 6 - 7 | 367 | 76 | |
AF252614.1 | - | 7 - 8 | 501 | 76 | |
AF257135.1 | - | 5 - 6 | 302 | 76 | |
AK002099.1 | - | 7 - 8 | 610 | 76 | |
AK092904.1 | - | 7 - 8 | 961 | 76 | |
AK290074.1 | - | 7 - 8 | 498 | 76 | |
AK290916.1 | - | 6 - 7 | 379 | 76 | |
AY190023.1 | - | 5 - 6 | 312 | 76 | |
BC001609.1 | - | 7 - 8 | 632 | 76 | |
BC009204.2 | - | 5 - 6 | 304 | 76 |