The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014146.3 | NP_054865.2 | 8 - 9 | 692 | 105 |
NM_032463.2 | NP_115852.1 | 9 - 10 | 811 | 105 | |
NM_032464.2 | NP_115853.2 | 9 - 10 | 881 | 105 | |
XM_011516558.2 | XP_011514860.1 | 9 - 10 | 721 | 105 | |
GenBank mRNA | AF161531.1 | - | 8 - 9 | 442 | 104 |
AF252611.1 | - | 9 - 10 | 632 | 105 | |
AF252613.1 | - | 8 - 9 | 428 | 105 | |
AF252614.1 | - | 9 - 10 | 562 | 105 | |
AF257135.1 | - | 7 - 8 | 363 | 105 | |
AK002099.1 | - | 9 - 10 | 671 | 105 | |
AK092904.1 | - | 9 - 10 | 1022 | 105 | |
AK225150.1 | - | 8 - 9 | 443 | 105 | |
AK290074.1 | - | 9 - 10 | 559 | 105 | |
AK290916.1 | - | 8 - 9 | 440 | 105 | |
AY190023.1 | - | 7 - 8 | 373 | 105 | |
BC001609.1 | - | 9 - 10 | 693 | 105 | |
BC009204.2 | - | 7 - 8 | 365 | 105 |