The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079812.2 | NP_001073280.1 | 22 - 23 | 3263 | 112 |
NM_001314007.1 | NP_001300936.1 | 23 - 24 | 3290 | 112 | |
NM_005219.4 | NP_005210.3 | 23 - 24 | 3290 | 112 | |
XM_011537572.2 | XP_011535874.1 | 22 - 23 | 3231 | 112 | |
XM_011537573.2 | XP_011535875.1 | 23 - 24 | 3098 | 112 | |
GenBank mRNA | AB209482.1 | - | 23 - 24 | 3231 | 112 |
AF051782.1 | - | 22 - 23 | 3077 | 112 | |
AK301195.1 | - | 8 - 9 | 1026 | 112 | |
AY363395.1 | - | 23 - 24 | 3149 | 112 | |
BC007411.2 | - | 7 - 8 | 701 | 112 | |
BC117257.1 | - | 22 - 23 | 3137 | 112 | |
BC143413.1 | - | 23 - 24 | 3167 | 112 | |
BC143414.1 | - | 23 - 24 | 3167 | 112 | |
BT007318.1 | - | 4 - 5 | 545 | 112 | |
DQ067453.1 | - | 22 - 23 | 3134 | 112 |