The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079812.2 | NP_001073280.1 | 5 - 6 | 730 | 85 |
NM_001314007.1 | NP_001300936.1 | 6 - 7 | 757 | 85 | |
NM_005219.4 | NP_005210.3 | 6 - 7 | 757 | 85 | |
XM_011537572.2 | XP_011535874.1 | 5 - 6 | 698 | 85 | |
XM_011537573.2 | XP_011535875.1 | 6 - 7 | 565 | 85 | |
GenBank mRNA | AB209482.1 | - | 6 - 7 | 698 | 85 |
AF051782.1 | - | 5 - 6 | 589 | 85 | |
AK307822.1 | - | 2 - 3 | 189 | 85 | |
AY363395.1 | - | 6 - 7 | 616 | 85 | |
BC117257.1 | - | 5 - 6 | 607 | 85 | |
BC143413.1 | - | 6 - 7 | 634 | 85 | |
BC143414.1 | - | 6 - 7 | 634 | 85 | |
BQ945734.1 | - | 6 - 7 | 691 | 85 | |
DQ067453.1 | - | 5 - 6 | 604 | 85 |