Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POF1B
Entrez Gene ID
79983
Gene Name
premature ovarian failure, 1B
Gene Aliases
POF, POF2B
Chromosome Location
Chr.X: 85277389 - 85379743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024921.3 NP_079197.3 16 - 17 1907 134
XM_005262203.3 XP_005262260.1 16 - 17 1820 134
GenBank mRNA AF309774.1 - 16 - 17 1907 135
AK025039.1 - 12 - 13 1500 134
AK025080.1 - 8 - 9 872 134
AK026445.1 - 4 - 5 400 134
AK225193.1 - 8 - 9 872 134
AK290360.1 - 16 - 17 1856 134

View Full Product Details