The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307940.1 | NP_001294869.1 | 2 - 3 | 426 | 140 |
NM_024921.3 | NP_079197.3 | 2 - 3 | 426 | 140 | |
XM_005262203.3 | XP_005262260.1 | 2 - 3 | 384 | 140 | |
GenBank mRNA | AF309774.1 | - | 2 - 3 | 426 | 140 |
AK025039.1 | - | 2 - 3 | 375 | 140 | |
AK290360.1 | - | 2 - 3 | 375 | 140 | |
AK314295.1 | - | 2 - 3 | 345 | 140 | |
BC017500.1 | - | 2 - 3 | 400 | 140 |