Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POF1B
Entrez Gene ID
79983
Gene Name
premature ovarian failure, 1B
Gene Aliases
POF, POF2B
Chromosome Location
Chr.X: 85277389 - 85379743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307940.1 NP_001294869.1 7 - 8 1003 156
NM_024921.3 NP_079197.3 7 - 8 1003 156
XM_005262203.3 XP_005262260.1 7 - 8 916 156
GenBank mRNA AF309774.1 - 7 - 8 1003 156
AK290360.1 - 7 - 8 952 156
AK314295.1 - 7 - 8 922 156
BC017500.1 - 7 - 8 977 156

View Full Product Details