Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC25A
Entrez Gene ID
993
Gene Name
cell division cycle 25A
Gene Aliases
CDC25A2
Chromosome Location
Chr.3: 48157178 - 48188658 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001789.2 NP_001780.2 12 - 13 1561 120
NM_201567.1 NP_963861.1 11 - 12 1441 120
XM_006713434.2 XP_006713497.1 12 - 13 1653 120
XM_006713435.2 XP_006713498.1 11 - 12 1533 120
XM_011534316.1 XP_011532618.1 13 - 14 1754 120
GenBank mRNA AK290554.1 - 12 - 13 1236 120
AY137580.1 - 11 - 12 1077 120
BC007401.2 - 12 - 13 1561 120
BC018642.2 - 12 - 13 1561 120
EU812118.1 - 11 - 12 1074 120
FJ866789.1 - 12 - 13 1194 120
M81933.1 - 12 - 13 1653 120

View Full Product Details