Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEB
Entrez Gene ID
4703
Gene Name
nebulin
Gene Aliases
NEB177D, NEM2
Chromosome Location
Chr.2: 151485334 - 151734487 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164507.1 NP_001157979.1 173 - 174 24685 66
NM_001164508.1 NP_001157980.1 173 - 174 24685 66
NM_001271208.1 NP_001258137.1 174 - 175 24790 66
NM_004543.4 NP_004534.2 141 - 142 19117 66
XM_005246590.2 XP_005246647.1 172 - 173 24579 66
XM_005246591.2 XP_005246648.1 172 - 173 24579 66
XM_005246592.2 XP_005246649.1 172 - 173 24579 66
XM_005246593.2 XP_005246650.1 173 - 174 24672 66
XM_005246594.2 XP_005246651.1 172 - 173 24579 66
XM_005246596.2 XP_005246653.1 171 - 172 24486 66
XM_005246598.2 XP_005246655.1 173 - 174 24672 66
XM_005246599.2 XP_005246656.1 170 - 171 24393 66
XM_005246600.2 XP_005246657.1 170 - 171 24393 66
XM_005246601.2 XP_005246658.1 169 - 170 24300 66
XM_005246602.2 XP_005246659.1 169 - 170 24300 66
XM_005246603.2 XP_005246660.1 168 - 169 24207 66
XM_005246604.2 XP_005246661.1 169 - 170 24300 66
XM_005246606.2 XP_005246663.1 168 - 169 24207 66
XM_005246612.2 XP_005246669.1 169 - 170 23943 66
XM_005246613.2 XP_005246670.1 170 - 171 23943 66
XM_005246617.2 XP_005246674.1 157 - 158 21756 66
XM_006712541.2 XP_006712604.1 168 - 169 24207 66
XM_011511226.2 XP_011509528.1 162 - 163 22485 66
XM_011511227.2 XP_011509529.1 149 - 150 20298 66
XM_017004177.1 XP_016859666.1 172 - 173 24561 66
XM_017004178.1 XP_016859667.1 171 - 172 24486 66
XM_017004179.1 XP_016859668.1 168 - 169 24207 66
XM_017004180.1 XP_016859669.1 168 - 169 24207 66
XM_017004181.1 XP_016859670.1 167 - 168 24114 66
XM_017004182.1 XP_016859671.1 167 - 168 24114 66
XM_017004184.1 XP_016859673.1 167 - 168 24114 66
GenBank mRNA AK056994.1 - 6 - 7 542 66
AK096887.1 - 1 - 2 81 66
AK300079.1 - 5 - 6 421 66
AK300674.1 - 4 - 5 376 66
AK300722.1 - 5 - 6 456 66
BC063136.1 - 6 - 7 523 66
X83957.1 - 128 - 129 19354 66

View Full Product Details