Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SEC61A2
- Entrez Gene ID
-
55176
- Gene Name
- Sec61 translocon alpha 2 subunit
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 12129641 - 12169958 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024576.2 |
|
12 - 13 |
1536 |
85 |
NR_024577.2 |
|
13 - 14 |
1669 |
85 |
GenBank mRNA |
AK057532.1 |
- |
13 - 14 |
1610 |
85 |
BC026179.1 |
- |
12 - 13 |
1486 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 548ak |
MIR548AK |
|
-36.9kb
|
Anti-sense |
Chr. - 12130760 - 12130816 on Build GRCh38 |
Downstream |
cell division cycle 123 |
CDC123 |
C10orf7, D123 |
26.0kb
|
Sense |
Chr. - 12195962 - 12250590 on Build GRCh38 |
View Full Product Details