Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SEC61A2
- Entrez Gene ID
-
55176
- Gene Name
- Sec61 translocon alpha 2 subunit
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 12129641 - 12169958 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024577.2 |
|
3 - 4 |
283 |
128 |
GenBank mRNA |
AK057532.1 |
- |
3 - 4 |
224 |
128 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
FY213180.1 |
- |
3 - 4 |
|
|
Chr. 10 - 12118780 - 12149667 [-] on Build GRCh38 |
128 |
GenBankmRNA |
DB454249.1 |
- |
3 - 4 |
256 |
|
Chr. 10 - 12129668 - 12143195 [+] on Build GRCh38 |
128 |
GenBankmRNA |
DB457301.1 |
- |
3 - 4 |
247 |
|
Chr. 10 - 12129677 - 12143196 [+] on Build GRCh38 |
128 |
GenBankmRNA |
DB064687.1 |
- |
3 - 4 |
222 |
|
Chr. 10 - 12129702 - 12149696 [+] on Build GRCh38 |
128 |
GenBankmRNA |
DB026597.1 |
- |
3 - 4 |
224 |
|
Chr. 10 - 12129701 - 12149713 [+] on Build GRCh38 |
128 |
GenBankmRNA |
HY040326.1 |
- |
3 - 4 |
205 |
|
Chr. 10 - 12129719 - 12143198 [+] on Build GRCh38 |
128 |
GenBankmRNA |
BP251468.1 |
- |
3 - 4 |
236 |
|
Chr. 10 - 12129688 - 12149728 [+] on Build GRCh38 |
128 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 548ak |
MIR548AK |
|
-5.3kb
|
Anti-sense |
Chr. - 12130760 - 12130816 on Build GRCh38 |
Downstream |
nudix hydrolase 5 |
NUDT5 |
YSA1, YSA1H, YSAH1 |
22.8kb
|
Anti-sense |
Chr. - 12165328 - 12196144 on Build GRCh38 |
View Full Product Details