The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017619.3 | NP_060089.1 | 3 - 4 | 475 | 123 |
GenBank mRNA | AK057799.1 | - | 3 - 4 | 454 | 123 |
AK289844.1 | - | 3 - 4 | 740 | 123 | |
AY099329.1 | - | 3 - 4 | 460 | 123 | |
BC010697.1 | - | 3 - 4 | 451 | 123 |