The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152573.3 | NP_689786.2 | 11 - 12 | 1834 | 125 |
XM_005251730.3 | XP_005251787.1 | 10 - 11 | 1814 | 125 | |
XM_005251731.3 | XP_005251788.1 | 11 - 12 | 2325 | 125 | |
GenBank mRNA | AK290986.1 | - | 11 - 12 | 1576 | 125 |
BC023566.1 | - | 11 - 12 | 1834 | 125 |