The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039465.1 | NP_001034554.1 | 3 - 4 | 476 | 69 |
NM_001320214.1 | NP_001307143.1 | 3 - 4 | 376 | 69 | |
NM_006925.4 | NP_008856.2 | 3 - 4 | 372 | 69 | |
XM_005267999.1 | XP_005268056.1 | 3 - 4 | 368 | 69 | |
XM_005268000.1 | XP_005268057.1 | 3 - 4 | 348 | 69 | |
XM_017021593.1 | XP_016877082.1 | 3 - 4 | 446 | 69 | |
GenBank mRNA | AB209804.1 | - | 3 - 4 | 291 | 69 |
AB451250.1 | - | 2 - 3 | 188 | 69 | |
AB451374.1 | - | 2 - 3 | 188 | 69 | |
AK296113.1 | - | 3 - 4 | 318 | 69 | |
AK300560.1 | - | 3 - 4 | 320 | 69 | |
AK311805.1 | - | 3 - 4 | 351 | 69 | |
BC018823.2 | - | 3 - 4 | 434 | 69 | |
BC040209.1 | - | 4 - 5 | 1642 | 69 | |
BF213008.1 | - | - | 352 | 69 | |
BT007089.1 | - | 2 - 3 | 188 | 69 | |
BU928782.1 | - | 3 - 4 | 479 | 69 | |
BX247988.1 | - | 3 - 4 | 416 | 69 | |
BX407495.2 | - | 3 - 4 | 301 | 69 | |
BX640605.1 | - | 3 - 4 | 371 | 69 | |
CR542167.1 | - | 2 - 3 | 188 | 69 | |
CR542182.1 | - | 2 - 3 | 188 | 69 | |
DC307911.1 | - | 3 - 4 | 348 | 69 | |
DR761538.1 | - | 3 - 4 | 304 | 69 | |
U30826.1 | - | 3 - 4 | 277 | 69 | |
U30827.1 | - | 3 - 4 | 406 | 69 | |
U30884.1 | - | 3 - 4 | 406 | 69 |