Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAC9
Entrez Gene ID
219348
Gene Name
placenta specific 9
Gene Aliases
-
Chromosome Location
Chr.10: 80132502 - 80145028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012973.1 NP_001012991.1 1 - 2 328 103
GenBank mRNA BC066348.1 - 1 - 2 328 103
BC090922.1 - 1 - 2 328 103
BC117332.1 - 1 - 2 156 103
BC117334.1 - 1 - 2 156 103

View Full Product Details