Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERBB3
Entrez Gene ID
2065
Gene Name
erb-b2 receptor tyrosine kinase 3
Gene Aliases
ErbB-3, HER3, LCCS2, MDA-BF-1, c-erbB-3, c-erbB3, erbB3-S, p180-ErbB3, p45-sErbB3, p85-sErbB3
Chromosome Location
Chr.12: 56080025 - 56103507 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001982.3 NP_001973.2 5 - 6 887 88
GenBank mRNA AK291681.1 - 5 - 6 740 88
AK294719.1 - 5 - 6 744 88
AK295650.1 - 6 - 7 654 88
BC002706.2 - 5 - 6 707 88
BC082992.1 - 5 - 6 726 88
BT007226.1 - 5 - 6 611 88
M29366.1 - 5 - 6 710 88
M34309.1 - 5 - 6 809 88

View Full Product Details