Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYYR1
Entrez Gene ID
116159
Gene Name
cysteine and tyrosine rich 1
Gene Aliases
C21orf95
Chromosome Location
Chr.21: 26466209 - 26573404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052954.3 NP_443186.2 3 - 4 805 152
GenBank mRNA AF401639.1 - 3 - 4 410 152
AK054581.1 - 3 - 4 685 152
AL833200.1 - 3 - 4 616 152
AY061853.1 - 3 - 4 350 152
BC036761.1 - 3 - 4 593 152
KF154022.1 - 4 - 5 577 152

View Full Product Details