The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113178.2 | NP_001106649.1 | 5 - 6 | 1346 | 64 |
NM_006599.3 | NP_006590.1 | 4 - 5 | 1292 | 64 | |
NM_138713.3 | NP_619727.2 | 5 - 6 | 1346 | 64 | |
NM_138714.3 | NP_619728.2 | 6 - 7 | 1408 | 64 | |
NM_173214.2 | NP_775321.1 | 5 - 6 | 1354 | 64 | |
NM_173215.2 | NP_775322.1 | 5 - 6 | 1354 | 64 | |
XM_006721125.3 | XP_006721188.1 | 4 - 5 | 2017 | 64 | |
XM_011522817.2 | XP_011521119.1 | 4 - 5 | 1292 | 64 | |
XM_011522818.2 | XP_011521120.1 | 7 - 8 | 1481 | 64 | |
XM_011522819.2 | XP_011521121.1 | 7 - 8 | 1063 | 64 | |
XM_011522820.2 | XP_011521122.1 | 4 - 5 | 493 | 64 | |
XM_011522822.2 | XP_011521124.1 | 3 - 4 | 441 | 64 | |
XM_017022870.1 | XP_016878359.1 | 4 - 5 | 1641 | 64 | |
XM_017022871.1 | XP_016878360.1 | 5 - 6 | 1368 | 64 | |
XM_017022872.1 | XP_016878361.1 | 4 - 5 | 1222 | 64 | |
XM_017022873.1 | XP_016878362.1 | 4 - 5 | 834 | 64 | |
GenBank mRNA | AB020634.1 | - | 4 - 5 | 1274 | 64 |
AF089824.1 | - | 5 - 6 | 1296 | 64 | |
AF134870.1 | - | 2 - 3 | 728 | 64 | |
AF163836.1 | - | 3 - 4 | 815 | 64 | |
AF346509.1 | - | 6 - 7 | 2294 | 64 | |
AJ005683.1 | - | 5 - 6 | 970 | 64 | |
AJ005684.1 | - | 5 - 6 | 970 | 64 | |
AJ243298.1 | - | 5 - 6 | 1113 | 64 | |
AJ243299.1 | - | 5 - 6 | 1105 | 64 | |
BC131509.1 | - | 5 - 6 | 1250 | 64 | |
BC146765.1 | - | 4 - 5 | 1274 | 64 |