The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145128.2 | NP_001138600.2 | 11 - 12 | 1149 | 145 |
NM_145025.4 | NP_659462.1 | 11 - 12 | 1149 | 145 | |
XM_006715376.3 | XP_006715439.2 | 12 - 13 | 2014 | 145 | |
XM_011535550.2 | XP_011533852.1 | 12 - 13 | 2014 | 145 | |
XM_011535552.2 | XP_011533854.1 | 11 - 12 | 1190 | 145 | |
XM_011535553.2 | XP_011533855.1 | 11 - 12 | 1797 | 145 | |
XM_011535555.2 | XP_011533857.1 | 12 - 13 | 2014 | 145 | |
XM_011535556.2 | XP_011533858.1 | 12 - 13 | 2014 | 145 | |
XM_011535559.2 | XP_011533861.1 | 12 - 13 | 2014 | 145 | |
XM_017010384.1 | XP_016865873.1 | 12 - 13 | 2014 | 145 | |
XM_017010385.1 | XP_016865874.1 | 11 - 12 | 1182 | 145 | |
XM_017010386.1 | XP_016865875.1 | 12 - 13 | 2014 | 145 | |
XM_017010387.1 | XP_016865876.1 | 6 - 7 | 735 | 145 | |
XM_017010388.1 | XP_016865877.1 | 12 - 13 | 2014 | 145 | |
GenBank mRNA | AK055538.1 | - | 11 - 12 | 1126 | 145 |
AK315561.1 | - | 11 - 12 | 1120 | 145 | |
AL832162.1 | - | 11 - 12 | 1128 | 145 | |
BC022031.2 | - | 11 - 12 | 1154 | 145 |